Learn More
Julabo™ Stainless Steel/PTFE External Pt100 Sensor
Brand:  Julabo™ 8981016
 View more versions of this product
Product Code. 11990268
  • 915.00 € / Each
Estimated Shipment: 15-10-2024
to see stock.
Explore more special offers
Add to basket
Specifications
External Pt100 Sensor
Suitable for CF31, CF41, F25-HE, F25-HL, F25-ME, F26-ME Refrigerated/Heating Circulator
PTFE-coated Stainless Steel
900 x 6 mm