Learn More
Sampling Systems™ Stainless Steel PharmaScoop™
Constructed of high quality Stainless Steel for durability. Sampling Systems™ Stainless Steel PharmaScoop™ is ideal for dispensing and sampling in pharmaceutical and hygienic environments.
54.45 € - 162.00 €
Specifications
Product Type | Powder Scoop |
---|---|
Material | Stainless Steel (316L) |
Finish | Mirror Polish |
Product Code | Brand | Capacity (Metric) | Length (Metric) | Length (Metric) Bowl | Diameter (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Length (Metric) Bowl | Diameter (Metric) | Price | Quantity & Availability | ||
12947955
|
Sampling Systems™
A316-1700 |
1.7 L | 355 mm | 230 mm | 135 mm |
133.00 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
12735018
|
Sampling Systems™
A316-100 |
100 mL | 180 mm | 100 mm | 50 mm |
73.95 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
12745018
|
Sampling Systems™
A316-500 |
500 mL | 240 mm | 140 mm | 95 mm |
98.00 €
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
||
12937955
|
Sampling Systems™
A316-1250 |
1250 mL | 305 mm | 195 mm | 125 mm |
121.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
15571792
|
Sampling Systems™
A316-1000 |
1000 mL | 340 mm | 235 mm | 110 mm |
108.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
12957955
|
Sampling Systems™
A316-750 |
750 mL | 330 mm | 210 mm | 95 mm |
106.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
15311481
|
Sampling Systems™
A316-50 |
50 mL | 110 mm | 70 mm | 38 mm |
55.95 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
15809621
|
Sampling Systems™
A316-10 |
10 mL | 75 mm | 45 mm | 25 mm |
54.45 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||
15819621
|
Sampling Systems™
A316-250 |
250 mL | 235 mm | 120 mm | 75 mm |
93.90 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
15829621
|
Sampling Systems™
A316-2500 |
2.5 L | 320 mm | 215 mm | 185 mm |
162.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
15839621
|
Sampling Systems™
A316-5000 |
5 L | 355 mm | 230 mm | 220 mm |
159.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||
Description
- Very high quality scoops
- Heavy duty construction
- Made from 316L Stainless Steel
- Crevice free
- Satin finish
- Rounded corners for easy cleaning
- GMP correct
Specifications
Powder Scoop | |
Stainless Steel (316L) | |
Mirror Polish |