missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Measuring Scoop

Ideal for scooping out fixed volumes of powder. Sampling Systems™ Stainless Steel Measuring Scoop is constructed of high quality Stainless Steel for durability.

Brand:  Sampling Systems™ 1900A-01

 View more versions of this product

Product Code. 15829301

  • 65.65 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured in 316L Stainless Steel
  • Accurately measures powders
Specifications

Specifications

Measuring Spoon
9mm Scoop Diameter and 14mm Scoop Depth
186 mm
Stainless Steel (316L)
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers