Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
673.00 € - 2660.00 €
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
15585129
|
Fisherbrand™
FB67849 |
1 g to 5 kg |
2660.00 €
1 unit |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
15515109
|
Fisherbrand™
FB67822 |
1 to 100 g |
673.00 €
1 unit |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
15505129
|
Fisherbrand™
FB67841 |
1 g to 1 kg |
1335.00 €
1 unit |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
Description
- Class F1 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
F1 | |
Cylindrical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F1