missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12822-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human SRRM1.
Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3′-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.Specifications
SRRM1 | |
Polyclonal | |
Unconjugated | |
SRRM1 | |
160 KD, POP101, SRM160, SRRM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10250, 313620, 51796 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q52KI8, Q8IYB3 | |
Srrm1 | |
SRRM1 Fusion Protein Ag3513 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction