missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Square Stainless Steel PharmaScoop™
Sampling Systems™ Square Stainless Steel PharmaScoop™ is suitable for hygienic and pharmaceutical applications.
Brand: Sampling Systems™ A366-1
This item is not returnable.
View return policy
Description
- Very high quality scoops
- Heavy duty construction
- Made from 316L Stainless Steel
- Crevice free
- Satin finish
- Rounded corners for easy cleaning
- GMP correct
Specifications
Stainless Steel (316L) | |
1500 mL | |
Mirror Polish |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction