missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15376-1-AP-150UL
This item is not returnable.
View return policy
Description
The IRF (interferon regulatory factor) family of transcription factors are characterized by an unique tryptophan pentad repeat DNA-binding domain. The IRFs are important in the regulation of interferons in response to infection by virus, and in the regulation of interferon-inducible genes. IRF4 is lymphocyte specific and negatively regulates toll-like-receptor (TLR) signaling that is central to the activation of innate and adaptive immune systems. A chromosomal translocation involving this gene and the IgH locus, t(6;14)(p25;q32), may be a cause of multiple myeloma. Alternatively spliced transcript variants have been found.Specifications
SPTLC1 | |
Polyclonal | |
Unconjugated | |
SPTLC1 | |
HSAN, HSAN1, HSN1, LBC1, LCB 1, LCB1, Serine palmitoyltransferase 1, SPT 1, SPT1, SPTI, SPTLC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10558, 268656, 361213 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15269, O35704 | |
Sptlc1 | |
SPTLC1 Fusion Protein Ag1162 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction