missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPPL2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13255-1-AP-150UL
This item is not returnable.
View return policy
Description
By searching sequence databases for homologs of Dictyostelium discoideum Impas, which shares homology with presenilin, followed by PCR of lymphocyte and hippocampus cDNA libraries, Grigorenko et al. (2002) cloned SPPL2a. This protein, which may act as an intramembrane protease, contains an N-terminal protease-associated (PA) domain, several transmembrane regions, a hydrophilic loop, conservative sequences around the first and second aspartate residues, and an invariant PAL motif near the C terminus.Specifications
SPPL2A | |
Polyclonal | |
Unconjugated | |
SPPL2A | |
IMP 3, IMP3, Intramembrane protease 3, Presenilin like protein 2, PSL2, SPP like 2A, SPPL2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311401, 66552, 84888 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TCT8, Q9JJF9 | |
Sppl2a | |
SPPL2A Fusion Protein Ag3935 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction