Learn More
Justrite™ SpillSlope™ Steel Shelf for 22 Gallon Undercounter Safety Cabinet
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 22 Gallon Undercounter Safety Cabinet adds additional storage to cabinets.
Brand: Justrite™ 29939
This item is not returnable.
View return policy
Description
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump. They adjust on 3 in. (76-mm) centers for versatile storage
- Fits 22 gal. Undercounter cabinet
Specifications
18 in. | |
Steel | |
31.56 in. |