missing translation for 'onlineSavingsMsg'
Learn More

SPI1 Mouse anti-Human, Clone: 2H3D3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66618-2-IG-150UL

 View more versions of this product

Product Code. 16818665

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

SPI1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17947
SPI1
Peptide
150 μL
Primary
Human
Antibody
IgG1
Western Blot
2H3D3
Unconjugated
SPI1
31 kDa transforming protein, OF, PU.1, SFPI1, SPI 1, SPI A, SPI1, Transcription factor PU.1
Mouse
Protein G
RUO
6688
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers