missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sphingosine-1-phosphate

158.79 € - 677.37 €

Chemical Identifiers

CAS 26993-30-6
Molecular Formula C18H38NO5P
Synonym D-erythro-Sphingosine 1-phosphate; S1P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15447829
SDS View Documents
Thermo Scientific Chemicals
J66459.M^
1 mg
158.79 €
1mg
Please to purchase this item. Need a web account? Register with us today!
15437829
SDS View Documents
Thermo Scientific Chemicals
J66459.#0
5 mg
677.37 €
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26993-30-6
C18H38NO5P
D-erythro-Sphingosine 1-phosphate; S1P

Specifications

White to off-white
26993-30-6
379.47
Light Sensitive
Powder
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
379.48
-30°C to -10°C
C18H38NO5P
D-erythro-Sphingosine 1-phosphate; S1P
Sphingosine-1-phosphate
DUYSYHSSBDVJSM-KRWOKUGFSA-N
{[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sphingosine-1-phosphate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.