missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermine tetrahydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
Solubility
Soluble in water (348.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.
Spermine tetrahydrochloride, is used to precipitate DNA from low salt aqueous buffers. Spermine tetrahydrochloride was used to precipitate DNA for flow cytometric analysis from fresh frozen breast cancer tissue, archival tissue of salivary gland, renal and thyroid tumors2 and red blood cells of chicken.
Solubility
Soluble in water (348.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away from strong oxidizing agents.
Specifications
Specifications
Color | White |
Quantity | 25 g |
Beilstein | 3911771 |
Solubility Information | Soluble in water (348.2g/L at 20°C). |
Formula Weight | 348.19 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | Spermine tetrahydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'