missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, 99%, Thermo Scientific Chemicals

Product Code. 11459396
Click to view available options
Quantity:
1 g
5 g
25 g
100 g

Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
PubChem CID 1102
ChEBI CHEBI:16610
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine
SMILES NCCCCNCCCN

Specifications

Melting Point 22°C to 25°C
Density 0.925
Boiling Point 128°C to 130°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 25 g
UN Number UN2735
Beilstein 1698591
Sensitivity Air sensitive
Merck Index 14,8742
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 145.25
Percent Purity 99%
Chemical Name or Material Spermidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'