missing translation for 'onlineSavingsMsg'
Learn More

Bellingham Stanley Spectral lamp outfit 110V 60Hz Bellingham and Stanley

Brand:  Bellingham Stanley 59-25

Spectral lamp outfit 110V 60Hz Bellingham and Stanley

Code : 04

Additional Details : Weight : 0.01000kg

Product Code. 12426230

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers