missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectinomycin dihydrochloride, 96.4%, MP Biomedicals™
112.00 € - 463.00 €
Chemical Identifiers
CAS | 22189-32-8 |
---|---|
Molecular Formula | C14H36Cl2N2O12 |
Molecular Weight (g/mol) | 495.34 |
MDL Number | MFCD00150886 |
InChI Key | DCHJOVNPPSBWHK-UXXUFHFZSA-N |
Synonym | U18409AE, Actinospectacin |
PubChem CID | 67108398 |
ChEBI | CHEBI:9217 |
IUPAC Name | dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride |
SMILES | [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
Description
Chemical Identifiers
22189-32-8 | |
495.34 | |
DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
67108398 | |
dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride |
C14H36Cl2N2O12 | |
MFCD00150886 | |
U18409AE, Actinospectacin | |
CHEBI:9217 | |
[H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
Specifications
Spectinomycin Dihydrochloride | |
22189-32-8 | |
C14H36Cl2N2O12 | |
U18409AE, Actinospectacin | |
[H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O | |
495.34 | |
CHEBI:9217 | |
Spectinomycin dihydrochloride |
Crystalline Powder | |
0.964 | |
MFCD00150886 | |
DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride | |
67108398 | |
495.36 |