Learn More
3M™ Aura™ Specialty Respirator, Series 9300
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand:  3M™ 7000034737
 View more versions of this product
Product Code. 11763373
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Fits a wide range of face shapes and sizes
  • Low breathing resistance filter technology for easier breathing
  • Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
  • An embossed top panel to reduce fogging of eyewear
  • A sculpted nose panel for improved compatibility with eyewear
  • An innovative chin tab for ease of donning and adjustment of the respirator on the face
  • Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
  • Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yellow
Mask FFP1 Acidic Gas
FFP1
Protection against dust, mist, and nuisnace level acid gas
9915
20/Pk.
Blue, Black
No
Electret (polyester, polypropylene, and carbon)
Cool-flow valve, color coded straps
Acid gas