missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18373-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein may play a role in fertility regulation. It were significantly upregulated in individual bile samples from Cholangiocarcinoma patients by Western blotting and Immunohistochemistry, relative to normal tissues. It acts as a potential serum diagnostic biomarker for Cholangiocarcinoma.Specifications
SPATA20 | |
Polyclonal | |
Unconjugated | |
SPATA20 | |
SPATA20, Sperm specific protein 411, spermatogenesis associated 20, SSP411, Tisp78 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217116, 360604, 64847 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6T393, Q80YT5, Q8TB22 | |
Spata20 | |
SPATA20 Fusion Protein Ag13234 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction