Learn More
BRAND™ Spare Lateral Stopcock with Key
Stopcock housing with burette tip includes interchangeable PTFE or borosilicate glass key with retention device.
Brand: BRAND™ 81142
This item is not returnable.
View return policy
Description
- Lateral stopcock housing with burette tip
- Interchangeable PTFE or borosilicate glass key with retention device
Specifications
50 mL | |
Borosilicate Glass | |
Retention Device |
Burette | |
1 Each | |
Stopcock Key |