missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Two-Color Wash Bottles - Safety-Vented™ and Safety Labeled, Wide-Mouth

Product Code. 15160386
Click to view available options
Capacity (Metric):
500 mL
Chemical Label:
Custom Write-on
Ethanol
Isopropanol
Methyl Ethyl Ketone
This item is not returnable. View return policy
Use only with solvents compatible with LDPE.
This item is not returnable. View return policy
Use only with solvents compatible with LDPE.

A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles

Volatiles are vented eliminating solvent drips. They are pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.

  • Valve eliminates drips; allows dispensing of wash bottle in an upright or inverted position
  • Wide mouth 53mm (2 in.) bottle for easy refilling
  • Polypropylene closure with leakproof design; color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

Order Info

8 oz. (250mL)

Specifications

Capacity 16 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 16 oz.
Color Translucent
Diameter (Metric) Outer Neck 53mm
For Use With (Application) Isopropanol
Includes Yellow Polypropylene Cap
Material Polyethylene
Quantity 1Each
Closure Color Yellow
Certifications/Compliance Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Autoclavable Not Autoclavable
Mouth Wide
Product Type Wash Bottle
Chemical Label Isopropanol
Closure Material Polypropylene
Closure Type Safety Vented Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'