missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ SP Scienceware™ Calibrated Electronic Stainless Steel Stem Thermometers: -50°/+200°C (-58°/+392°F)

Choose from a variety of features in these digital stem thermometers that are suitable for field studies, wet lab applications, wash down areas, food testing and tanks.

Brand:  H-B Instrument™ B60900-2000

 View more versions of this product

Product Code. 17520936

  • 55.05 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

These electronic stem thermometers are not suitable for solids such as frozen items


Includes: Multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)

Requires : LR44 battery
This item is not returnable. View return policy

Description

Description

  • Thermometers provide a range of −50°/200°C (−58°/392°F);13-201-768 is −40°/232°C (−40°/450°F)
  • Resolution of 0.1°C/°F (13-201-770 is 1°C/°F)
  • Will read temperatures in liquids, semi-solids and gases
  • 6.7 in. to 10.75 in. probes easily reach into cuvettes, test tubes, flasks, beakers and more
  • Blunt tip probe is ideal for veterinary practices, farm and ranch
  • Surface probe is ideal for cooking, laboratories and hot plates (does not include probe cover)
  • Features include minimum/maximum temperature readings, alarms, data hold, water resistant models and more

Certifications

CE marked

Compliance

Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST; individually serialized

Specifications

Specifications

0°, 100°C
190 x 20 mm (7.5 x 0.8 in.)
Stainless Steel
0.1°C/°F
200°C
-50°C to +200°C
±1°C from –20°/120°C; otherwise ±2°C
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No.2448.01), against equipment whose calibration is traceable to NIST. Individually serialized, CE marked.
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Stainless Steel
Auto Off; 120mm (4.7 in.) Stainless Steel Probe; Requires LR44 battery
-50°C
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers