missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Acetal Plastic Screw Clamp

Bel-Art™ SP Scienceware™ Acetal Plastic Screw Clamp offers on/off and fine control so accurate that it replaces stopcocks in many applications.

Brand:  Bel-Art™ 182120000

Product Code. 10172391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Can be installed without disconnecting tubing or interrupting operation
  • Accepts flexible tubing up to 14mm (1/2 in.) O.D.
  • Molded of autoclavable acetal plastic
  • Resistant to acids, alkalis and organic solvents
Specifications

Specifications

Autoclavable
Screw Style Tubing Clamp
Fits Tubing O.D. 0.5 in. (1.27cm) in.
0 to 0.5 in.
0 to 1.27 cm
3
Acetal Plastic
2.31 in.
5.867 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.