Learn More
3M™ Solus™ Safety Glasses
Click to view available options
Lens Tint:
Amber
Blue, Mirror
Bronze
Clear
Indoor/Outdoor, Mirror
Red, Mirror
Lens Coating:
Scratch-resistant
Description
- Lens curve has been calculated to optimize the fitting, field of vision and coverage of the eyes
- Extremely stylish and lightweight glasses
- Soft inner padding of the temple, leading to increased comfort
- Supplied with a microfiber storage bag for cleaning and protection
- Anti-scratch and anti-fog coating for increased durability and improved vision
Specifications
Specifications
Certifications/Compliance | CE Approved, EN 166: 2001 |
Lens Tint | Indoor/Outdoor, Mirror |
Product Type | Safety glasses |
Frame Color | Black, Orange |
Includes | Microfiber storage bag |
Lens Material | Polycarbonate |