missing translation for 'onlineSavingsMsg'
Learn More

Sodium trifluoroacetate, 98%, Thermo Scientific Chemicals

38.29 € - 510.54 €

Chemical Identifiers

CAS 2923-18-4
Molecular Formula C2F3NaO2
Molecular Weight (g/mol) 136.01
MDL Number MFCD00013217
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
PubChem CID 517019
SMILES [Na+].[O-]C(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11452048
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14613.14
25 g
38.29 €
25g
Please to purchase this item. Need a web account? Register with us today!
11462048
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14613.22
100 g
115.58 €
100g
Please to purchase this item. Need a web account? Register with us today!
11472048
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14613.36
500 g
510.54 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Specifications

Chemical Identifiers

2923-18-4
136.01
UYCAUPASBSROMS-UHFFFAOYSA-M
517019
C2F3NaO2
MFCD00013217
sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
[Na+].[O-]C(=O)C(F)(F)F

Specifications

2923-18-4
98%
MFCD00013217
Hygroscopic
Soluble in water.
[Na+].[O-]C(=O)C(F)(F)F
136.01
136
Sodium Trifluoroacetate
∽207°C (decomposition)
C2F3NaO2
3597949
sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
UYCAUPASBSROMS-UHFFFAOYSA-M
sodium;2,2,2-trifluoroacetate
517019
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 220-879-6

missing translation for 'rtecsNumber' : AK0250000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' : Hygroscopic

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium trifluoroacetate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H300-Fatal if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : Sodium trifluoroacetate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only