missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide trihydrate, 96%, Thermo Scientific Chemicals

37.59 € - 402.00 €

Chemical Identifiers

CAS 12209-98-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.732
MDL Number MFCD00150189
InChI Key SFXJSNATBHJIDS-UHFFFAOYSA-N
Synonym sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
PubChem CID 21977542
IUPAC Name disodium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488527
View Documents
Thermo Scientific Alfa Aesar
A18611.22
100 g
37.59 €
100g
Please to purchase this item. Need a web account? Register with us today!
11498527
View Documents
Thermo Scientific Alfa Aesar
A18611.36
500 g
108.54 €
500g
Please to purchase this item. Need a web account? Register with us today!
11408537
View Documents
Thermo Scientific Alfa Aesar
A18611.0E
2500 g
402.00 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

12209-98-2
266.732
SFXJSNATBHJIDS-UHFFFAOYSA-N
21977542
O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]
H6Na2O6Sn
MFCD00150189
sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
disodium;dioxido(oxo)tin;trihydrate

Specifications

12209-98-2
MFCD00150189
sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
SFXJSNATBHJIDS-UHFFFAOYSA-N
disodium;dioxido(oxo)tin;trihydrate
21977542
98%
Sodium tin(IV) oxide trihydrate
H6Na2O6Sn
14,8677
Soluble in water. Insoluble in ethanol and acetone.
O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]
266.732
266.75 (212.67 Anhydrous)
Odorless
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 235-030-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.