missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulphate 0.1 mol/L (0.1 N) in aqueous solution, BAKER ANALYZED™, J.T. Baker™
Laboratory Reagent
106.00 € - 155.00 €
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | Sodium hyposulphite , di-Sodium thiosulphate |
IUPAC Name | disodium pentahydrate sulfanidesulfonate |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10526622
|
JT Baker
7101.1000 |
1 L |
106.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10698331
|
JT Baker
7101.9010 |
10 L |
155.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate |
Na2O3S2 | |
MFCD00003499 | |
Sodium hyposulphite , di-Sodium thiosulphate | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
7772-98-7 | |
MFCD00003499 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate | |
0.0997 - 0.1003 mol/L |
Na2O3S2 | |
Sodium hyposulphite , di-Sodium thiosulphate | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
Plastic bottle |
Safety and Handling
Recommended Storage : Ambient
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title