missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Pentahydrate, puriss., Honeywell Fluka™
Puriss., meets analytical specification of BP, European Pharmacopoeia, USP, 99-101%
26.36 € - 444.56 €
Chemical Identifiers
CAS | 10102-17-7 |
---|---|
Molecular Formula | H10Na2O8S2 |
Molecular Weight (g/mol) | 248.172 |
MDL Number | MFCD00149186 |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15651350
|
Honeywell-Fluka
13479-500G |
500 g |
26.36 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15631350
|
Honeywell-Fluka
13479-2.5KG |
2.5 kg |
86.69 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15661350
|
Honeywell-Fluka
13479-5KG |
5 kg |
172.25 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15671350
|
Honeywell-Fluka
13479-6X2.5KG |
6 x 2.5 kg | |||||||
15641350
|
Honeywell-Fluka
13479-4X5KG |
4 x 5 kg | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10102-17-7 | |
248.172 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
H10Na2O8S2 | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Specifications
10102-17-7 | |
Na2S2O3 · 5H2O | |
NONH for all modes of transport | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.18g/mol | |
puriss. |
H10Na2O8S2 | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 | |
99 to 101% | |
Sodium Thiosulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling
missing translation for 'einecsNumber' : 231-867-5