missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, Honeywell
Purum p.a., anhydrous,≥98.0% (RT)
22.51 € - 73.54 €
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
PubChem CID | 24477 |
IUPAC Name | disodium pentahydrate sulfanidesulfonate |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
24477 | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
disodium pentahydrate sulfanidesulfonate |
Specifications
7772-98-7 | |
Na2S2O3 | |
NONH for all modes of transport | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate | |
24477 | |
≥98% (RT) |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.11g/mol | |
Sodium thiosulfate |
Safety and Handling
missing translation for 'rtecsNumber' : XN6476000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title