missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate concentrate, For 1L standard solution, 0.1 M Na2S2O3 (0.1N), Honeywell Fluka™

for 1L standard solution, 0.1 M Na2S2O3 (0.1N)

32.42 € - 201.18 €

Chemical Identifiers

CAS 10102-17-7, 6132-02-1, 7732-18-5
MDL Number MFCD00003499
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15645240
View Documents
Honeywell-Fluka
38200-1EA
1 Pc.
32.42 €
Each
Please to purchase this item. Need a web account? Register with us today!
15655240
View Documents
Honeywell Fluka
38200-6X1EA
6 x 1 Ea.
201.18 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10102-17-7, 6132-02-1, 7732-18-5
MFCD00003499

Specifications

10102-17-7, 6132-02-1, 7732-18-5
Na2S2O3
NONH for all modes of transport
AKHNMLFCWUSKQB-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
24477
Ampoule
Na2O3S2
MFCD00003499
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
[Na+].[Na+].[O-]S([S-])(=O)=O
158.10
158.11g/mol
Sodium thiosulfate concentrate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.