missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
21.41 € - 155.78 €
Chemical Identifiers
CAS | 7772-98-7 |
---|---|
Molecular Formula | Na2O3S2 |
Molecular Weight (g/mol) | 158.10 |
MDL Number | MFCD00003499 |
InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
PubChem CID | 24477 |
SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15213406
|
Thermo Scientific Acros
450622500 |
250 g |
21.41 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15271906
|
Thermo Scientific Acros
450620010 |
1 kg |
43.11 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15219656
|
Thermo Scientific Acros
450620050 |
5 kg |
155.78 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
24477 |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Specifications
48.0°C | |
6.0 to 8.4 (5% aq. soln.) | |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
24477 | |
99% | |
1% max. (105°C) |
7772-98-7 | |
98.5% min. (Iodimetry) | |
Na2S2O3 | |
15,8821 | |
Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.1 | |
Analytical | |
Sodium thiosulfate |
Safety and Handling
missing translation for 'einecsNumber' : 231-867-5
RUO – Research Use Only