Learn More
Sodium thiocyanate, 98%, extra pure, Thermo Scientific Chemicals
34.80 € - 132.00 €
Chemical Identifiers
CAS | 540-72-7 |
---|---|
Molecular Formula | CNNaS |
Molecular Weight (g/mol) | 81.07 |
MDL Number | MFCD00011123 |
InChI Key | VGTPCRGMBIAPIM-UHFFFAOYSA-M |
Synonym | sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased |
PubChem CID | 516871 |
ChEBI | CHEBI:30952 |
IUPAC Name | sodium;thiocyanate |
SMILES | [Na+].[S-]C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10572655
|
Thermo Scientific Acros
196762500 |
250 g | Plastic bottle |
34.80 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266300
|
Thermo Scientific Acros
196765000 |
500 g | Plastic bottle |
65.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10379133
|
Thermo Scientific Acros
196760025 |
2.5 kg | Plastic bottle |
132.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
540-72-7 | |
81.07 | |
VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
516871 | |
sodium;thiocyanate |
CNNaS | |
MFCD00011123 | |
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
CHEBI:30952 | |
[Na+].[S-]C#N |
Specifications
287.0°C | |
White | |
Crystalline Powder, Crystals and/or Chunks | |
CNNaS | |
MFCD00011123 | |
15, 9480 | |
Solubility in water: 1390g/L (21°C). Other solubilities: soluble in alcohol, freely soluble in acetone | |
[Na+].[S-]C#N | |
81.07 | |
CHEBI:30952 | |
98% | |
2.5% max. |
540-72-7 | |
5.5 to 7.5 (5% soln. at 20°C) | |
97.5% min. (on dried substance) (Argentometry) | |
NaSCN | |
01,1105 | |
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased | |
VGTPCRGMBIAPIM-UHFFFAOYSA-M | |
sodium;thiocyanate | |
516871 | |
81.07 | |
Extra Pure | |
Sodium thiocyanate, 98% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Contact with acids liberates very toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 208-754-4
missing translation for 'rtecsNumber' : XL2275000
missing translation for 'tsca' : TSCA
- Sodium thiocyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P273-Avoid release to the environment.
- P233-Keep container tightly closed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only