missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, ACS Reagent, Anhydrous Granular, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
12 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Physical Form | Granules |
Quantity | 12 kg |
Linear Formula | Na2SO4 |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium;sulfate |
Formula Weight | 142.04g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Density | 2.68g/mL at 25°C |
Chemical Name or Material | Sodium Sulfate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'