missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate Solution, Reagent Grade, Honeywell™

Product Code. 15618370
Click to view available options
Quantity:
25 mL
1 L
3 L
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Quantity 25 mL
Linear Formula Na2O(SiO2)x · xH2O
UN Number UN3266
Packaging Poly bottle
Density 1.39g/mL at 25°C
Chemical Name or Material Sodium silicate solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'