Learn More
Sodium pyrophosphate decahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
31.36 € - 181.91 €
Chemical Identifiers
CAS | 13472-36-1 |
---|---|
Molecular Formula | H20Na4O17P2 |
Molecular Weight (g/mol) | 446.05 |
MDL Number | MFCD00149200 |
InChI Key | VZWGHDYJGOMEKT-UHFFFAOYSA-J |
Synonym | sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate |
PubChem CID | 3084150 |
IUPAC Name | tetrasodium;phosphonato phosphate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337718
|
Thermo Scientific Alfa Aesar
033385.22 |
100 g |
31.36 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327718
|
Thermo Scientific Alfa Aesar
033385.36 |
500 g |
55.48 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317718
|
Thermo Scientific Alfa Aesar
033385.A3 |
2 kg |
181.91 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13472-36-1 | |
446.05 | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
3084150 | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] |
H20Na4O17P2 | |
MFCD00149200 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
tetrasodium;phosphonato phosphate;decahydrate |
Specifications
79.5°C | |
White | |
99-103% | |
MFCD00149200 | |
Soluble in water. Insoluble in alcohol and ammonia. | |
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+] | |
446.05 | |
446.06 (265.90 Anhydrous) | |
White crystalline | |
Sodium pyrophosphate decahydrate, For analysis ACS |
13472-36-1 | |
Crystalline | |
H20Na4O17P2 | |
sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate | |
VZWGHDYJGOMEKT-UHFFFAOYSA-J | |
tetrasodium;phosphonato phosphate;decahydrate | |
3084150 | |
ACS | |
1.82 g/cm3 at 20°C |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 231-767-1
missing translation for 'rtecsNumber' : UX7350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only