Learn More
Sodium phosphate, tribasic dodecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
35.40 € - 110.00 €
Chemical Identifiers
CAS | 10101-89-0 |
---|---|
Molecular Formula | Na3O4P·12H2O |
Molecular Weight (g/mol) | 380.13 |
MDL Number | MFCD00149198 |
InChI Key | ASTWEMOBIXQPPV-UHFFFAOYSA-K |
Synonym | trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate |
PubChem CID | 61473 |
IUPAC Name | trisodium;phosphate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10158190
|
Thermo Scientific Acros
424405000 |
500 g | Plastic bottle |
35.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10582364
|
Thermo Scientific Acros
424400025 |
2.5 kg | Plastic bottle |
110.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10101-89-0 | |
380.13 | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
61473 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] |
Na3O4P·12H2O | |
MFCD00149198 | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
trisodium;phosphate;dodecahydrate |
Specifications
75.0°C | |
White | |
98+% | |
Na3PO4·12H2O | |
15, 8792 | |
Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+] | |
380.13 | |
380.13 | |
ACS Reagent | |
1.6200g/mL |
10101-89-0 | |
Crystalline Powder or Needles | |
Na3O4P·12H2O | |
MFCD00149198 | |
trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate | |
ASTWEMOBIXQPPV-UHFFFAOYSA-K | |
trisodium;phosphate;dodecahydrate | |
61473 | |
≥98% | |
1.62 | |
Sodium phosphate, tribasic dodecahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : TC9575000
- Sodium phosphate, tribasic dodecahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Tribasic sodium phosphate dodecahydrate
RUO – Research Use Only