missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals), Fisher BioReagents™
26.10 € - 41.95 €
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 139.00 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
SMILES | O.[Na+].OP(O)(O)=O |
Description
Sodium Phosphate Monobasic Monohydrate (Colorless-to-white Crystals)
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O |
Specifications
100°C | |
White | |
Solid | |
MFCD00149208 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
98.0 to 102.0% | |
Sodium Phosphate Monobasic Monohydrate |
10049-21-5 | |
4.1 to 4.5 | |
H5NaO5P | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
Pass Test |
Safety and Handling
Recommended Storage : RT