missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, monobasic monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Sodium phosphate, monobasic monohydrate, 98%, H2NaO4P.H2O, CAS Number-10049-21-5
72.26 € - 311.55 €
Chemical Identifiers
CAS | 10049-21-5 |
---|---|
Molecular Formula | H5NaO5P |
Molecular Weight (g/mol) | 139.00 |
MDL Number | MFCD00149208 |
InChI Key | BBMHARZCALWXSL-UHFFFAOYSA-N |
Synonym | sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 |
PubChem CID | 516949 |
SMILES | O.[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12695097
|
Thermo Scientific Acros
447740010 |
1 kg |
72.26 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12605107
|
Thermo Scientific Acros
447740050 |
5 kg |
311.55 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10049-21-5 | |
139.00 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
516949 |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
O.[Na+].OP(O)(O)=O |
Specifications
100.0°C | |
98% | |
NaH2PO4·H2O | |
15,879 | |
Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol | |
O.[Na+].OP(O)(O)=O | |
139.00 | |
137.99 | |
Extra Pure | |
Sodium phosphate, monobasic monohydrate |
10049-21-5 | |
H5NaO5P | |
MFCD00149208 | |
sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707 | |
BBMHARZCALWXSL-UHFFFAOYSA-N | |
sodium;dihydrogen phosphate;hydrate | |
516949 | |
98% | |
Plastic bottle |
Safety and Handling
Product Identifier
- Sodium phosphate, monobasic monohydrate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title