missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate Monobasic Anhydrous (Colorless-to-White Crystals), Fisher BioReagents™
For tissue culture and molecular biology applications
57.20 € - 85.75 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
PubChem CID | 23672064 |
ChEBI | CHEBI:37585 |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10133153
|
Fisher BioReagents
BP329-500 |
500 g | Glass Bottle |
57.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173103
|
Fisher BioReagents
BP329-1 |
1 kg | Poly Bottle |
85.75 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For tissue culture and molecular biology applications
Sodium Phosphate is commonly used in tissue culture and molecular biology applications.Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
[Na+].OP(O)([O-])=O |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
CHEBI:37585 |
Specifications
203.9°C | |
7558-80-7 | |
4.1 to 4.5 | |
≥99 % | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
≥99% | |
2.04g/cm³ |
100°C | |
White | |
Solid | |
H2NaO4P | |
15, 8790 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
119.98 | |
Pass Test | |
Sodium Phosphate Monobasic Anhydrous |
Safety and Handling
Recommended Storage : RT