missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 4 x 2.5 kg |
Linear Formula | Na2HPO4 |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium;hydrogen phosphate |
Formula Weight | 141.96g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium phosphate dibasic |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'