missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium persulfate, Honeywell
Purum p.a.,≥99.0% (RT)
46.02 € - 245.82 €
Chemical Identifiers
CAS | 7775-27-1 |
---|---|
Molecular Formula | Na2O8S2 |
Molecular Weight (g/mol) | 238.092 |
MDL Number | MFCD00003501 |
InChI Key | CHQMHPLRPQMAMX-UHFFFAOYSA-L |
Synonym | sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate |
PubChem CID | 62655 |
IUPAC Name | disodium;sulfonatooxy sulfate |
SMILES | [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15656860
|
Honeywell Research Chemicals
71890-500G |
500 g |
46.02 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15646860
|
Honeywell Research Chemicals
71890-1KG |
1 kg |
60.91 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15666860
|
Honeywell Research Chemicals
71890-5KG |
5 kg |
245.82 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7775-27-1 | |
238.092 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
62655 | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
disodium;sulfonatooxy sulfate |
Specifications
7775-27-1 | |
Na2S2O8 | |
UN1505 | |
CHQMHPLRPQMAMX-UHFFFAOYSA-L | |
disodium;sulfonatooxy sulfate | |
62655 | |
≥99% (RT) |
Na2O8S2 | |
MFCD00003501 | |
sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate | |
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+] | |
238.092 | |
238.1g/mol | |
Sodium persulfate |
Safety and Handling
P210-P220-P261-P280-P284-P370 + P378
missing translation for 'rtecsNumber' : SE0525000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium persulfate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.