missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, Reagent grade, Honeywell™

Product Code. 15602210
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, granular, +140 mesh particle size, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Physical Form Granules
Quantity 500 g
UN Number 1504
Formula Weight 77.98g/mol
Percent Purity 97%
Grade Reagent
Chemical Name or Material Sodium Peroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'