Learn More
Sodium peroxide, 96%, Thermo Scientific Chemicals
38.20 € - 336.00 €
Chemical Identifiers
CAS | 1313-60-6 |
---|---|
Molecular Formula | Na2O2 |
Molecular Weight (g/mol) | 77.97 |
MDL Number | MFCD00003497 |
InChI Key | PFUVRDFDKPNGAV-UHFFFAOYSA-N |
Synonym | sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide |
PubChem CID | 14803 |
IUPAC Name | disodium;peroxide |
SMILES | [O-][O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10412021
|
Thermo Scientific Acros
207700250 |
25 g | Plastic bottle |
38.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174880
|
Thermo Scientific Acros
207701000 |
100 g | Plastic bottle |
93.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626792
|
Thermo Scientific Acros
207705000 |
500 g | Plastic bottle |
336.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-60-6 | |
77.97 | |
PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
14803 | |
[O-][O-].[Na+].[Na+] |
Na2O2 | |
MFCD00003497 | |
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
disodium;peroxide |
Specifications
460.0°C | |
Yellow | |
96% | |
Na2O2 | |
06,548; 07,341; 11,492 | |
sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide | |
PFUVRDFDKPNGAV-UHFFFAOYSA-N | |
disodium;peroxide | |
14803 | |
96% | |
2.8000g/mL |
1313-60-6 | |
Granules | |
Na2O2 | |
MFCD00003497 | |
15, 8786 | |
Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2, freely soluble | |
[O-][O-].[Na+].[Na+] | |
77.97 | |
77.97 | |
2.8 | |
Sodium peroxide, p.a. |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-209-4
missing translation for 'tsca' : TSCA
- Sodium peroxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only