missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Periodate, Certified AR for Analysis, Fisher Chemical™
INaO4, CAS Number-7790-28-5, sodiumperiodate, sodium m-periodate, periodate sodium, sodium periodate, periodic acid hio4 , sodium salt, sodium meta periodate, sodium penodate, sodium metaperiodate, periodic acid, sodium salt, sodium meta-periodate, 250g, CHEBI:75226, White, 213.89g/mol
85.70 € - 1195.00 €
Chemical Identifiers
CAS | 7790-28-5 |
---|---|
Molecular Formula | INaO4 |
Molecular Weight (g/mol) | 213.89 |
MDL Number | MFCD00003534 |
InChI Key | JQWHASGSAFIOCM-UHFFFAOYSA-M |
Synonym | sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate |
PubChem CID | 23667635 |
ChEBI | CHEBI:75226 |
IUPAC Name | sodium;periodate |
SMILES | [Na+].[O-][I](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418962
|
Fisher Chemical
S/5980/48 |
100 g |
85.70 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11443423
|
Fisher Chemical
S/5980/50 |
250 g |
161.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413753
|
Fisher Chemical
S/5980/65 |
5 kg |
1195.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INaO4, CAS Number-7790-28-5, sodiumperiodate, sodium m-periodate, periodate sodium, sodium periodate, periodic acid hio4 , sodium salt, sodium meta periodate, sodium penodate, sodium metaperiodate, periodic acid, sodium salt, sodium meta-periodate, 250g, CHEBI:75226, White, 213.89g/mol
Chemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
sodium;periodate |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 | |
[Na+].[O-][I](=O)(=O)=O |
Specifications
300°C | |
White | |
INaO4 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
Plastic powder jar |
7790-28-5 | |
3.4 to 4.5 | |
MFCD00003534 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium;periodate | |
23667635 | |
213.89g/mol |