Learn More
Sodium periodate, 99.8+%, ACS reagent, Thermo Scientific Chemicals
24.20 € - 1355.00 €
Chemical Identifiers
CAS | 7790-28-5 |
---|---|
Molecular Formula | INaO4 |
Molecular Weight (g/mol) | 213.89 |
MDL Number | MFCD00003534 |
InChI Key | JQWHASGSAFIOCM-UHFFFAOYSA-M |
Synonym | sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate |
PubChem CID | 23667635 |
ChEBI | CHEBI:75226 |
SMILES | [Na+].[O-][I](=O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10647391
|
Thermo Scientific Acros
419610050 |
5 g | Glass Bottle |
24.20 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348673
|
Thermo Scientific Acros
419611000 |
100 g | Plastic bottle |
96.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229990
|
Thermo Scientific Acros
419615000 |
500 g | Plastic bottle |
340.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924221
|
Thermo Scientific Acros
419610025 |
2.5 kg | Plastic Bottle |
1355.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7790-28-5 | |
213.89 | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
23667635 | |
[Na+].[O-][I](=O)(=O)=O |
INaO4 | |
MFCD00003534 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
CHEBI:75226 |
Specifications
300.0°C | |
White | |
99.8 to 100.3% (on dried basis) (Iodometry) | |
NaIO4 | |
01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294 | |
sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate | |
JQWHASGSAFIOCM-UHFFFAOYSA-M | |
sodium;periodate | |
23667635 | |
213.89 | |
ACS Reagent |
7790-28-5 | |
Crystalline Powder | |
INaO4 | |
MFCD00003534 | |
15,8772 | |
Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents | |
[Na+].[O-][I](=O)(=O)=O | |
213.89 | |
CHEBI:75226 | |
≥99.8% | |
Sodium periodate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Very toxic to aquatic life.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-197-6
- Sodium periodate
Signal Word
- Danger
Hazard Category
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only