missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perchlorate monohydrate, 85+%, ACS reagent, Thermo Scientific Chemicals
25.43 € - 246.23 €
Chemical Identifiers
CAS | 7791-07-3 |
---|---|
InChI Key | IXGNPUSUVRTQGW-UHFFFAOYSA-M |
Synonym | sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate |
PubChem CID | 516899 |
IUPAC Name | sodium;perchlorate;hydrate |
SMILES | O.[O-]Cl(=O)(=O)=O.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10757002
|
Thermo Scientific Acros
343631000 |
100 g | Plastic bottle |
25.43 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306691
|
Thermo Scientific Acros
343630010 |
1 kg | Plastic bottle |
113.57 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914221
|
Thermo Scientific Acros
343630025 |
2.5 kg | Plastic bottle |
246.23 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7791-07-3 | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
sodium;perchlorate;hydrate |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
516899 | |
O.[O-]Cl(=O)(=O)=O.[Na+] |
Specifications
130.0°C | |
White to Yellow | |
Crystalline Powder | |
ClNaO4·H2O | |
15,8784 | |
Solubility in water: 2090g/L (15°C). Other solubilities: soluble in alcohol | |
O.[O-]Cl(=O)(=O)=O.[Na+] | |
140.46 | |
140.46 | |
ACS Reagent |
7791-07-3 | |
6.0 to 8.0 (5% aq. soln. at 25°C) | |
85.0 to 90% (as NaClO4) (Argentometry) | |
ClNaO4·H2O | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
sodium;perchlorate;hydrate | |
516899 | |
≥85% | |
Sodium perchlorate monohydrate |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
Harmful if swallowed.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium perchlorate monohydrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only