missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%
35.65 € - 150.22 €
Chemical Identifiers
CAS | 7632-00-0 |
---|---|
Molecular Formula | NNaO2 |
Molecular Weight (g/mol) | 69.00 |
MDL Number | MFCD00011118 |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
PubChem CID | 23668193 |
ChEBI | CHEBI:78870 |
IUPAC Name | sodium;nitrite |
SMILES | [Na+].[O-]N=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633430
|
Honeywell-Fluka
31443-100G |
100 g |
35.65 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15653430
|
Honeywell-Fluka
31443-250G |
250 g |
60.09 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15663430
|
Honeywell-Fluka
31443-500G |
500 g |
77.60 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643430
|
Honeywell-Fluka
31443-1KG |
1 kg |
150.22 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7632-00-0 | |
69.00 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
23668193 | |
sodium;nitrite |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
CHEBI:78870 | |
[Na+].[O-]N=O |
Specifications
7632-00-0 | |
NaNO2 | |
1500 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
sodium;nitrite | |
23668193 | |
69g/mol | |
ACS/puriss. p.a. | |
Sodium Nitrite, Reag. Ph. Eur. |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
[Na+].[O-]N=O | |
69.00 | |
CHEBI:78870 | |
≥99% | |
914°F |
Safety and Handling
P220-P273-P301 + P310-P305 + P351 + P338
H272-H301-H319-H400
missing translation for 'einecsNumber' : 231-555-9
missing translation for 'rtecsNumber' : RA1225000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- sodium nitrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium nitrate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title