Learn More
Thermo Scientific Chemicals Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
83.82 € - 288.44 €
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15460767
|
Thermo Scientific Chemicals
J67606.22 |
100 g |
83.82 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15470767
|
Thermo Scientific Chemicals
J67606.30 |
250 g |
186.93 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480767
|
Thermo Scientific Chemicals
J67606.36 |
500 g |
288.44 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
CHEBI:8984 |
Spezifikation
151-21-3 | |
White | |
MFCD00036175 | |
3599286 | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38 | |
Molecular Biology | |
Sodium n-dodecyl sulfate |
∼206°C | |
C12H25NaO4S | |
UN2926 | |
14,8636 | |
Soluble in water | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
99% | |
Solid |
Sicherheit und Handhabung
GHS H Statement
H228-H311-H302-H315-H319-H335
Flammable solid.
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H332-H315-H318-H335-H500
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 205-788-1
missing translation for 'rtecsNumber' : WT1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Signalwort
- Danger
Gefahrenkategorie
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Ergänzende Informationen
- MIXTURE LIST-Contains : Sodium lauryl sulfate
RUO – Research Use Only