missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-dodecyl sulfate, 97%, for electrophoresis, Thermo Scientific Chemicals

Product Code. 15426279
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

Solubility
Soluble in water. Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Melting Point ∼206°C
Quantity 100 g
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
Merck Index 14,8636
Solubility Information Soluble in water; Slightly soluble in ethanol.
IUPAC Name sodium;dodecyl sulfate
Formula Weight 288.38
Percent Purity 97%
Chemical Name or Material Sodium n-dodecyl sulfate
Grade Ultrapure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium n-dodecyl sulfate, ultrapure
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'