missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, puriss., Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 6 x 2.5 kg |
Linear Formula | Na2MoO4 · 2H2O |
UN Number | NONH for all modes of transport |
IUPAC Name | disodium dioxomolybdenumbis(olate) |
Formula Weight | 241.95 |
Percent Purity | 99 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Chemical Name or Material | Sodium Molybdate Dihydrate, Meets analytical specification of PhEur |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'