Learn More
Sodium metavanadate, typically 96%, V 38% min, Thermo Scientific Chemicals
24.92 € - 329.64 €
Chemical Identifiers
CAS | 13718-26-8 |
---|---|
Molecular Formula | NaO3V |
Molecular Weight (g/mol) | 121.93 |
MDL Number | MFCD00011125 |
InChI Key | CMZUMMUJMWNLFH-UHFFFAOYSA-N |
Synonym | sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881 |
PubChem CID | 4148882 |
ChEBI | CHEBI:75221 |
SMILES | [Na+].[O-][V](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336108
|
Thermo Scientific Alfa Aesar
011092.14 |
25 g |
24.92 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346108
|
Thermo Scientific Alfa Aesar
011092.30 |
250 g |
97.59 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356108
|
Thermo Scientific Alfa Aesar
011092.A1 |
1 kg |
329.64 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13718-26-8 | |
121.93 | |
CMZUMMUJMWNLFH-UHFFFAOYSA-N | |
4148882 | |
[Na+].[O-][V](=O)=O |
NaO3V | |
MFCD00011125 | |
sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881 | |
CHEBI:75221 |
Specifications
630°C | |
White | |
Typically 96% | |
MFCD00011125 | |
Soluble in water. | |
[Na+].[O-][V](=O)=O | |
121.93 | |
CHEBI:75221 | |
Odorless | |
Sodium metavanadate, V 38% min. |
13718-26-8 | |
Chunks and/or Powder | |
NaO3V | |
sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881 | |
CMZUMMUJMWNLFH-UHFFFAOYSA-N | |
sodium;oxido(dioxo)vanadium | |
4148882 | |
121.93 | |
Off-white to light orange powder |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c
H301-H319-H332-H361-H372-H500
missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 237-272-7
missing translation for 'rtecsNumber' : YW1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium metavanadate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium metavanadate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.