Learn More
Sodium metatungstate hydrate, crystalline, Thermo Scientific Chemicals
83.11 € - 682.40 €
Chemical Identifiers
CAS | 652154-12-6 |
---|---|
Molecular Formula | H2Na6O40W12·xH2O |
MDL Number | MFCD00150663 |
Synonym | Sodium polytungstate Hydrate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091601
|
Thermo Scientific Acros
304090500 |
50 g | Glass bottle |
83.11 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173281
|
Thermo Scientific Acros
304092500 |
250 g | Glass bottle |
281.40 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10423075
|
Thermo Scientific Acros
304090010 |
1 kg | Glass bottle |
682.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
652154-12-6 | |
H2Na6O40W12·xH2O | |
MFCD00150663 | |
Sodium polytungstate Hydrate |
Specifications
652154-12-6 | |
Crystalline Powder or Beads | |
Na6[H2W12O40]·xH2O | |
Sodium polytungstate Hydrate | |
2986.13 | |
Sodium metatungstate hydrate |
White to Yellow | |
H2Na6O40W12·xH2O | |
MFCD00150663 | |
Solubility in water: >1000g/L (20°C). | |
≥85% (WO3) |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes serious eye damage. Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment.
Danger
- Sodium metatungstate hydrate, crystalline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.